Learn More
AdipoGen TrisDEAcyclopropenylium DICY
SDP
Catalog No. 502560017
Encompass_Preferred
Chemical. CAS 1415962-26-3. Formula C17H30N6. MW 318.46. Synthetic Ionic liquid. The properties of these ionic liquids include almost zero vapour pressure, low flammability and ease of recycling. They are frequently considered as potential green alternatives to classical organic solvents. Additionally, most classes can be readily tuned to provide unique solvent environments in which to carry out reactions, extractions and other processes. There are a large number of applications.
Catalog No. 50-256-0017 Supplier AdipoGen Supplier No. CDXT0089G001
Chemical. CAS 1415962-26-3. Formula C17H30N6. MW 318.46. Synthetic Ionic liquid. The properties of these ionic liquids include almost zero vapour pressure, low flammability and ease of recycling. They are frequently considered as potential green alternatives to classical organic solvents. Additionally, most classes can be readily tuned to provide unique solvent environments in which to carry out reactions, extractions and other processes. There are a large number of applications.