Learn More
AdipoGen TrisDEAcyclopropenylium trif
SDP
Supplier:  AdipoGen CDXT0091G005
Encompass_Preferred
Chemical. CAS 1333477-58-9. Formula C17H30F6N4O4S2. MW 532.56. Synthetic Air- and water-stable ionic liquid. The properties of these ionic liquids include almost zero vapour pressure, low flammability and ease of recycling. They are frequently considered as potential green alternatives to classical organic solvents. Additionally, most classes can be readily tuned to provide unique solvent environments in which to carry out reactions, extractions and other processes. There are a large number of applications.
Catalog No. 50-256-0014