Learn More
AdipoGen Trishydroxymethyl aminometha
SDP
Catalog No. 502559958
Encompass_Preferred
Chemical. CAS 77-86-1. Formula C4H11NO3. MW 121.14. Synthetic. A high quality buffer used in the range of physiological pH 7-9. Widely used as a biological buffer or as a component of buffer formulations. Tris has a pKa of 8.06 and is very useful in the biology and biochemistry lab because it has buffering capabilities in the range of the typical physiological pH of most living organisms pH 7.0-9.0. Tris has been reported to interfere with the activity of a number of enzymes, it should therefore be used carefully when studying proteins.
Catalog No. 50-255-9958 Supplier AdipoGen Supplier No. CDXT0244G025
Chemical. CAS 77-86-1. Formula C4H11NO3. MW 121.14. Synthetic. A high quality buffer used in the range of physiological pH 7-9. Widely used as a biological buffer or as a component of buffer formulations. Tris has a pKa of 8.06 and is very useful in the biology and biochemistry lab because it has buffering capabilities in the range of the typical physiological pH of most living organisms pH 7.0-9.0. Tris has been reported to interfere with the activity of a number of enzymes, it should therefore be used carefully when studying proteins.