Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TrisHydroxymethyl)Aminomethane ACS AR, Macron Fine Chemicals™

Catalog No. MK1806100 Shop All Avantor Products
Change view
Click to view available options
Quantity:
100 g
500 g
5 kg
Packaging:
Glass
STAKMOR
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-1806-100 100 g Glass
MK-1806-500 500 g Glass
MK-1806-5 5 kg STAKMOR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MK-1806-100 Supplier Avantor Supplier No. 180657
Add to Cart
Add to Cart

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Quantity 100 g
Packaging Glass
Formula Weight 121.14
Grade ACS
Chemical Name or Material Tris(Hydroxymethyl)Aminomethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.