Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trismat(R) Hydrochloride, Crystal, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18614595 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
125 g
2.5 kg
5 kg
25 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-595 5 kg Poly Bottle
18-614-594 25 g Poly Bottle
18-614-590 125 g Poly Bottle
18-614-592 2.5 kg Poly Bottle
18-614-593 25 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-614-595 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SP3075KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1185-53-1
Molecular Weight (g/mol) 157.59

Specifications

pH 4.2 to 4.9
CAS Max % 1
Heavy Metals 5e-06
Packaging Poly Bottle
Quantity 5 kg
Physical Form Crystal
Percent Purity 99 to 101%
Grade Reagent
Chemical Name or Material Trismat Hydrochloride

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.