Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trisodium Citrate Anhydrous, DPL

Catalog No. 502985592
Encompass
Catalog No. 50-298-5592 Supplier Thermo Scientific Production Chemicals and Services Supplier No. D502010002
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

Molecular Weight (g/mol) 258.07 g/mol

Specifications

Boiling Point Undetermined
Quantity 25 kg
Percent Purity 99.0 to 100.5%
Grade FCC
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.