Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUJIFILM Biosciences Trisodium Citrate Dihydrate, 99.0% (Titration) Purity, 1 kg

Catalog No. NC3953031
Encompass_Preferred

Trisodium citrate dihydrate is a white crystalline reagent used to prepare sodium citrate buffers and saline-sodium citrate (SSC) solutions; it is supplied as a molecular biology grade chemical tested for DNase and RNase activities and suitable for laboratory buffer preparation and other molecular biology applications.

  • Molecular biology grade reagent
  • Purity 99.0+% (titration)
  • CAS number 6132-04-3
  • Molecular formula C6H5Na3O7·2H2O
  • Molecular weight 294.10 g·mol-1
  • Appearance white crystals or crystalline powder
  • Packaging 1 kg bottle
  • Storage store at room temperature

Catalog No. NC3953031 Supplier FUJIFILM Biosciences Supplier No. 20216671
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trisodium citrate dihydrate is a white crystalline reagent used to prepare sodium citrate buffers and saline-sodium citrate (SSC) solutions; it is supplied as a molecular biology grade chemical tested for DNase and RNase activities and suitable for laboratory buffer preparation and other molecular biology applications.

  • Molecular biology grade reagent
  • Purity 99.0+% (titration)
  • CAS number 6132-04-3
  • Molecular formula C6H5Na3O7·2H2O
  • Molecular weight 294.10 g·mol-1
  • Appearance white crystals or crystalline powder
  • Packaging 1 kg bottle
  • Storage store at room temperature

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.