Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trisodium citrate dihydrate, 99%

Catalog No. AAA122740E
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA122740E 2500 g
AAA1227436 500 g
AAA122740C 10,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA122740E Supplier Thermo Scientific Chemicals Supplier No. A122740E
Only null left
Add to Cart

CAS: 6132-04-3 | C6H9Na3O9 | 294.10 g/mol

Trisodium citrate dihydrate, is widely applied in food, beverages and fillers as a buffering, sequestering or an emulsifying agent. It used as an anticoagulant in blood transfusions, osmotic laxative, functional fluids, solvents cleaning, furnishing care products, laundry dishwashing products and cleaning automobile radiators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trisodium citrate dihydrate, is widely applied in food, beverages and fillers as a buffering, sequestering or an emulsifying agent. It used as an anticoagulant in blood transfusions, osmotic laxative, functional fluids, solvents cleaning, furnishing care products, laundry dishwashing products and cleaning automobile radiators.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Stable and incompatible with bases, reducing agents, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4-3-6132
Molecular Formula C6H9Na3O9
Molecular Weight (g/mol) 294.10
MDL Number MFCD00150031
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
Synonym trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
PubChem CID 71474
ChEBI CHEBI:32142
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]

Specifications

Melting Point 150°C (in -2H2O)
Density 1.76
Odor Odorless
Quantity 2500 g
Beilstein 6104939
Merck Index 14,8602
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 294.10 (258.07 Anhydrous)
Percent Purity 99%
Chemical Name or Material Trisodium citrate dihydrate
Hazard Category H500
EINECSNumber 200-675-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.