Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trisodium Paraperiodate 98.0+%, TCI America™
SDP

Catalog No. P007725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P007725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P007725G Supplier TCI America Supplier No. P007725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13940-38-0
Molecular Formula H2INa3O6
Molecular Weight (g/mol) 293.88
MDL Number MFCD00036146
InChI Key DSYMXPNUZLZKOA-UHFFFAOYSA-K
Synonym sodium paraperiodate, sodium para periodate, sodium triparaperiodate, sodium hydrogen periodate, 3na.h2io6, periodic acid trisodium salt, trisodium dihydrogen orthoperiodate, sodium para periodate, commercial, trisodium dihydroxydioxidoiodosylolate, trisodium dihydroxy-trioxido-oxo-$l^ 7-iodane
PubChem CID 9796123
IUPAC Name trisodium (dihydroxydioxidoiodosyl)olate
SMILES [Na+].[Na+].[Na+].O[I](O)([O-])([O-])([O-])=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 25 g
UN Number 1479
Formula Weight 293.88
Percent Purity ≥98.0% (T)
Chemical Name or Material Trisodium Paraperiodate
EINECSNumber (1)-0444
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.