Learn More
Medchemexpress LLC Trisodium phosphate | 7601-54-9 | MFCD00003510 | 99.5% | 163.94 g/mol | Na3PO4 | 100g
SDP
Catalog No. 5000266521
Encompass_Preferred
Trisodium phosphate is a food additive Trisodium phosphate affects the growth parameters (lag phase maximum growth rate) of Listeria monocytogenes and increases the sensitivity of Gram-negative bacteria to Lysozyme (HY-P1068) and Nisin (HY-P1607)[1][2]
Catalog No. 50-002-66521 Supplier Medchemexpress LLC Supplier No. HYY1329100G
Trisodium phosphate is a food additive Trisodium phosphate affects the growth parameters (lag phase maximum growth rate) of Listeria monocytogenes and increases the sensitivity of Gram-negative bacteria to Lysozyme (HY-P1068) and Nisin (HY-P1607)[1][2]