Learn More
Medchemexpress LLC Trisodium phosphate (dodecahydrate) | 10101-89-0 | 98.1% | 380.18 g/mol | Na3PO4.12H2O | 500 G
SDP
Catalog No. 5000208437
Encompass_Preferred
Trisodium phosphate dodecahydrate is an inorganic reagent used as a pH regulator, buffer, and corrosion inhibitor in laboratory and pharmaceutical applications. It is supplied as a white to off-white solid with high water content in the dodecahydrate form.
  • High purity reagent for analytical and preparative use.
  • Acts as a buffer and pH adjuster in aqueous solutions.
  • Used as a corrosion inhibitor in electrochemical studies.
  • Solid, white to off-white crystals suitable for handling and storage.
  • Available in multiple pack sizes including 500 g.
Catalog No. 50-002-08437 Supplier Medchemexpress LLC Supplier No. HYW088066500G
Trisodium phosphate dodecahydrate is an inorganic reagent used as a pH regulator, buffer, and corrosion inhibitor in laboratory and pharmaceutical applications. It is supplied as a white to off-white solid with high water content in the dodecahydrate form.
  • High purity reagent for analytical and preparative use.
  • Acts as a buffer and pH adjuster in aqueous solutions.
  • Used as a corrosion inhibitor in electrochemical studies.
  • Solid, white to off-white crystals suitable for handling and storage.
  • Available in multiple pack sizes including 500 g.