Learn More
Medchemexpress LLC Trisodium phosphate dodecahydrate | 10101-89-0 | MFCD00149198 | 98.1% | 380.12 g/mol | Na3PO4 · 12H2O | 1 KG
SDP
Catalog No. 5000208440
Encompass_Preferred
Trisodium phosphate dodecahydrate is an inorganic, research-grade salt used as a corrosion inhibitor, pH regulator, buffer component, and pharmaceutical excipient. It is provided as a crystalline hydrate with a listed assay of 98.1% and is offered in multiple pack sizes including 1 kg.
  • Corrosion inhibitor for iron in aqueous systems.
  • Functions as a pH regulator and buffer component.
  • Available in multiple pack sizes for laboratory use.
  • Supplied as a crystalline dodecahydrate (hydrated form).
Catalog No. 50-002-08440 Supplier Medchemexpress LLC Supplier No. HYW0880661KG
Trisodium phosphate dodecahydrate is an inorganic, research-grade salt used as a corrosion inhibitor, pH regulator, buffer component, and pharmaceutical excipient. It is provided as a crystalline hydrate with a listed assay of 98.1% and is offered in multiple pack sizes including 1 kg.
  • Corrosion inhibitor for iron in aqueous systems.
  • Functions as a pH regulator and buffer component.
  • Available in multiple pack sizes for laboratory use.
  • Supplied as a crystalline dodecahydrate (hydrated form).