Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trisodium phosphate dodecahydrate | 10101-89-0 | MFCD00149198 | 98.1% | 380.12 g/mol | Na3PO4·12H2O | 2.5 KG
SDP

Catalog No. 5000208436
Encompass_Preferred

Trisodium phosphate dodecahydrate is an inorganic salt supplied as a white to off-white crystalline solid, commonly used as a reagent, buffer component, pH modifier, and excipient for laboratory and pharmaceutical research applications.

  • Purity 98.1% assay.
  • Chemical formula Na3PO4·12H2O.
  • Molecular weight 380.12 g/mol.
  • Appearance white to off-white crystalline solid.
  • Soluble in water ≥ 100 mg/mL (≈263.07 mM).
  • Available in multiple pack sizes, including 2.5 kg.

Catalog No. 50-002-08436 Supplier Medchemexpress LLC Supplier No. HYW0880662.5KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trisodium phosphate dodecahydrate is an inorganic salt supplied as a white to off-white crystalline solid, commonly used as a reagent, buffer component, pH modifier, and excipient for laboratory and pharmaceutical research applications.

  • Purity 98.1% assay.
  • Chemical formula Na3PO4·12H2O.
  • Molecular weight 380.12 g/mol.
  • Appearance white to off-white crystalline solid.
  • Soluble in water ≥ 100 mg/mL (≈263.07 mM).
  • Available in multiple pack sizes, including 2.5 kg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.