Learn More
Medchemexpress LLC Trisodium phosphate (dodecahydrate) | 10101-89-0 | MFCD00149198 | 98.1% | 380.12 g/mol | Na3PO4·12H2O | 250 G
SDP
Catalog No. 5000208470
Encompass_Preferred
Trisodium phosphate dodecahydrate is an inorganic phosphate salt supplied as a white crystalline powder. It functions as a pH regulator, buffer component, pharmaceutical excipient, and corrosion inhibitor for iron in chloride solutions; listed CAS number is 10101-89-0 and listed purity is 98.1%.
  • High purity (98.1%) suitable for laboratory use.
  • Dodecahydrate form for predictable aqueous solubility and buffering.
  • Solid powder form simplifies weighing and formulation.
  • Useful as a pH modifier and buffer component in formulation work.
  • Available in small pack sizes for bench-scale experiments.
Catalog No. 50-002-08470 Supplier Medchemexpress LLC Supplier No. 250GHYW088066
Trisodium phosphate dodecahydrate is an inorganic phosphate salt supplied as a white crystalline powder. It functions as a pH regulator, buffer component, pharmaceutical excipient, and corrosion inhibitor for iron in chloride solutions; listed CAS number is 10101-89-0 and listed purity is 98.1%.
  • High purity (98.1%) suitable for laboratory use.
  • Dodecahydrate form for predictable aqueous solubility and buffering.
  • Solid powder form simplifies weighing and formulation.
  • Useful as a pH modifier and buffer component in formulation work.
  • Available in small pack sizes for bench-scale experiments.