Learn More
Medchemexpress LLC Tristearin-d105 | 125941-88-0 | 99.6% | 10 MG
SDP
Catalog No. 5000341314
Encompass_Preferred
Tristearin-d105 is a deuterium-labeled form of Tristearin, a triglyceride derived from three units of stearic acid. Deuterium labeling has the potential to affect the pharmacokinetic and metabolic profiles of drugs, making this compound suitable for use in various research applications.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only
  • An isotope-labeled compound
Catalog No. 50-003-41314 Supplier Medchemexpress LLC Supplier No. HY127035S10MG
Tristearin-d105 is a deuterium-labeled form of Tristearin, a triglyceride derived from three units of stearic acid. Deuterium labeling has the potential to affect the pharmacokinetic and metabolic profiles of drugs, making this compound suitable for use in various research applications.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only
  • An isotope-labeled compound