Learn More
Medchemexpress LLC Tristearin-d5 | 55256-03-6 | 98.9% | 25 MG
SDP
Catalog No. 5000342733
Encompass_Preferred
Tristearin-d5 is a deuterium-labeled Tristearin, a triglyceride derived from three units of stearic acid. This compound has several applications:
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-42733 Supplier Medchemexpress LLC Supplier No. 25MGHY127035S3
Tristearin-d5 is a deuterium-labeled Tristearin, a triglyceride derived from three units of stearic acid. This compound has several applications:
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.