Learn More
Medchemexpress LLC Tristearin-d5 | 55256-03-6 | 98.9% | 50 MG
SDP
Catalog No. 5000341568
Encompass_Preferred
Tristearin-d5 is a deuterium labeled triglyceride derived from three units of stearic acid. It is intended for research use only, not for patient use. Stable heavy isotopes, such as deuterium in Tristearin-d5, are often incorporated into drug molecules for use as tracers during the drug development process, with the potential to influence pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-41568 Supplier Medchemexpress LLC Supplier No. HY127035S350MG
Tristearin-d5 is a deuterium labeled triglyceride derived from three units of stearic acid. It is intended for research use only, not for patient use. Stable heavy isotopes, such as deuterium in Tristearin-d5, are often incorporated into drug molecules for use as tracers during the drug development process, with the potential to influence pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS