Learn More
Medchemexpress LLC Tristearin-d9 | 285979-76-2 | MFCD01075451 | >=98.0% | 900.54 g/mol | C57H101D9O6 | 5 MG
SDP
Catalog No. 5000263478
Encompass_Preferred
Tristearin-d9 is the deuterium-labeled form of tristearin, a triglyceride made from three stearic acid units. It is used as a stable isotope-labeled standard in analytical and metabolic research, supporting accurate quantitation and tracing in lipid studies.
  • Deuterium-labeled triglyceride for use as an internal standard in analytical methods.
  • High isotopic enrichment (≈99 atom % D) and ≥98% chemical purity for reliable quantitation.
  • Appropriate for mass spectrometry and lipid metabolism tracing studies.
  • Supplied in a research-scale quantity suitable for method development.
  • Stable under typical storage conditions; follow provided handling instructions and documentation.
Catalog No. 50-002-63478 Supplier Medchemexpress LLC Supplier No. HY127035S45MG
Tristearin-d9 is the deuterium-labeled form of tristearin, a triglyceride made from three stearic acid units. It is used as a stable isotope-labeled standard in analytical and metabolic research, supporting accurate quantitation and tracing in lipid studies.
  • Deuterium-labeled triglyceride for use as an internal standard in analytical methods.
  • High isotopic enrichment (≈99 atom % D) and ≥98% chemical purity for reliable quantitation.
  • Appropriate for mass spectrometry and lipid metabolism tracing studies.
  • Supplied in a research-scale quantity suitable for method development.
  • Stable under typical storage conditions; follow provided handling instructions and documentation.