Learn More
AdipoGen TRITC
SDP
Supplier:  AdipoGen CDXT0011M250
Encompass_Preferred
Chemical. CAS 95197-95-8. Formula C25H21N3O3S. MW 443.52. Synthetic. 56-TRITC is spectrally similar to TAMRA. The isothiocyanate reactive group reacts with primary or secondary amines to form a thiourea bond linkage.
Catalog No. 50-256-0048