Learn More
AdipoGen TRITC (50 mg)
SDP
Supplier:  AdipoGen CDXT0011M050
Encompass_Preferred
Chemical. CAS: 95197-95-8. Formula: C25H21N3O3S. MW: 443.52. Synthetic. 5(6)-TRITC is spectrally similar to TAMRA. The isothiocyanate reactive group reacts with primary or secondary amines to form a thiourea bond linkage.
Catalog No. 50-205-4653