Learn More
Chondrex Inc TRITC-DEXTRAN, 125 MG, 70 KDA
Catalog No. 501526775
Encompass_Preferred
TRITC-Dextran (70 kDa) solution, 25 mg/mL x 5 mL, in PBS (red color). Fluorescent Properties: Excitation - 550 nm, Emission - 572 nm.
Catalog No. 50-152-6775 Supplier Chondrex Inc Supplier No. 4014
TRITC-Dextran (70 kDa) solution, 25 mg/mL x 5 mL, in PBS (red color). Fluorescent Properties: Excitation - 550 nm, Emission - 572 nm.