Learn More
Cayman Chemical Triticonazole, Purity: ≥95%, CAS: 131983-72-7, 50 mg
SDP
Catalog No. 502966174
Encompass_Preferred
A triazole fungicide that inhibits ergosterol biosynthesis
Catalog No. 50-296-6174 Supplier Cayman Chemical Supplier No. 1872650 MG
A triazole fungicide that inhibits ergosterol biosynthesis