Learn More
Andwin Scientific TRITON 100, 50ML
SDP
Catalog No. NC9160286
Encompass_Preferred
Triton X-100, Protein Grade, Detergent 10 Solution 10 aqueous Solution. Packaged under inert gas, Contaminants aldehyde concentration <200mm peroxide concentration, <200mm conductivity <50mmhos, 1/EA
Catalog No. NC9160286 Supplier Andwin Scientific Supplier No. 648463
Triton X-100, Protein Grade, Detergent 10 Solution 10 aqueous Solution. Packaged under inert gas, Contaminants aldehyde concentration <200mm peroxide concentration, <200mm conductivity <50mmhos, 1/EA