Learn More
Bioworld Triton Lysis Buffer, pH 7.4, 500 mL
Catalog No. 50253064
Encompass_Preferred
Triton Lysis Buffer, 1X Lysis buffer for extracting proteins from cells. Technical Information:1X concentrationTris-HCl, pH 7.4 (50mM)NaCl (150mM)Triton X-100 (1%)EDTA (5mM)
Catalog No. 50-253-064 Supplier Bioworld Supplier No. 220400572
Triton Lysis Buffer, 1X Lysis buffer for extracting proteins from cells. Technical Information:1X concentrationTris-HCl, pH 7.4 (50mM)NaCl (150mM)Triton X-100 (1%)EDTA (5mM)