Learn More
Research Products International Corp Triton Lysis Buffer Solution, 500 ML
SDP
Catalog No. 501978244
Encompass_Preferred
1X buffer used to lyse cells under non-denaturing conditions.
1X Formulation:
TRIS HCI: (pH 7.4) 50 mM
NaCI: 150 mM
TritonX-100: 1%
EDTA: 5 mM
Catalog No. 50-197-8244 Supplier Research Products International Corp Supplier No. T81200500.0
1X buffer used to lyse cells under non-denaturing conditions.
1X Formulation:
TRIS HCI: (pH 7.4) 50 mM
NaCI: 150 mM
TritonX-100: 1%
EDTA: 5 mM