Learn More
Alkali Scientific Triton Lysis Buffer Solution, 1 L
SDP
Catalog No. 5000661819
Encompass_Preferred
Triton Lysis Buffer Solution is a powerful reagent used for efficient cell lysis and protein extraction in molecular biology and biochemistry applications. This 1L solution is optimized for breaking down cell membranes, enabling the release of intracellular proteins for downstream analysis. Triton X-100, a non-ionic detergent, is the primary active ingredient that solubilizes the lipid bilayer, making it ideal for applications that require protein extraction while maintaining protein integrity. The buffer's gentle formulation ensures that it does not degrade proteins, making it suitable for applications such as Western blotting, immunoprecipitation, and enzyme assays. This buffer is widely used in research for cell-based assays, protein studies, and various biochemical tests.
Catalog No. 50-006-61819 Supplier Alkali Scientific Supplier No. RIT001CP
Triton Lysis Buffer Solution is a powerful reagent used for efficient cell lysis and protein extraction in molecular biology and biochemistry applications. This 1L solution is optimized for breaking down cell membranes, enabling the release of intracellular proteins for downstream analysis. Triton X-100, a non-ionic detergent, is the primary active ingredient that solubilizes the lipid bilayer, making it ideal for applications that require protein extraction while maintaining protein integrity. The buffer's gentle formulation ensures that it does not degrade proteins, making it suitable for applications such as Western blotting, immunoprecipitation, and enzyme assays. This buffer is widely used in research for cell-based assays, protein studies, and various biochemical tests.