Learn More
MSC Triton Products Single Ring Tool Holder Pegboard Hook, 1/2 in ID, 3/16 in Dia, 1-1/8 in Long
Catalog No. 5000529175
Encompass
This single ring tool holder pegboard hook is designed for organizing tools on a pegboard. Constructed from steel with a zinc finish, it provides a durable solution for heavy items. The hook features a 1/2 in inside diameter, 3/16 in rod thickness, and a 1-1/8 in projection, making it suitable for various tools.
  • Hook type: single ring tool holder
  • Designed for heavy wire type
  • Mounts on pegboard
  • Made of steel
  • Zinc finish
  • Projection of 1-1/8 in
  • For peg hole diameter of 3/8 in
  • Silver color
  • Two mounting holes
  • Rod thickness of 3/16 in
  • Includes locking system
  • Hole spacing of 1-1/2 in
  • Mounting hardware included
  • Inside diameter of 1/2 in
  • Country of origin: China
Catalog No. 50-005-29175 Supplier MSC Supplier No. 03096492
This single ring tool holder pegboard hook is designed for organizing tools on a pegboard. Constructed from steel with a zinc finish, it provides a durable solution for heavy items. The hook features a 1/2 in inside diameter, 3/16 in rod thickness, and a 1-1/8 in projection, making it suitable for various tools.
  • Hook type: single ring tool holder
  • Designed for heavy wire type
  • Mounts on pegboard
  • Made of steel
  • Zinc finish
  • Projection of 1-1/8 in
  • For peg hole diameter of 3/8 in
  • Silver color
  • Two mounting holes
  • Rod thickness of 3/16 in
  • Includes locking system
  • Hole spacing of 1-1/2 in
  • Mounting hardware included
  • Inside diameter of 1/2 in
  • Country of origin: China