Learn More
MSC Triton Products U-Shape Pegboard Hook, 2-7/8 in L, 1-3/4 in I.D., Zinc Plated Steel, 5 Pack
Catalog No. 5000532957
Encompass
The U-Shaped Hook is a double mount pegboard hook designed to securely lock onto pegboards for heavy-duty holding. Constructed from durable zinc plated steel, it provides robust and reliable storage for various tools, including those with long handles.
  • Heavy-duty holding capability for long handle tools.
  • Securely locks onto pegboard on both sides.
  • Dimensions of 2-7/8 in length and 1-3/4 in inside diameter.
  • Constructed from zinc plated steel for durability.
  • Includes a locking system for added security.
  • Supplied with mounting hardware for easy installation.
  • Comes in a convenient pack of 5.
Catalog No. 50-005-32957 Supplier MSC Supplier No. 03096799
The U-Shaped Hook is a double mount pegboard hook designed to securely lock onto pegboards for heavy-duty holding. Constructed from durable zinc plated steel, it provides robust and reliable storage for various tools, including those with long handles.
  • Heavy-duty holding capability for long handle tools.
  • Securely locks onto pegboard on both sides.
  • Dimensions of 2-7/8 in length and 1-3/4 in inside diameter.
  • Constructed from zinc plated steel for durability.
  • Includes a locking system for added security.
  • Supplied with mounting hardware for easy installation.
  • Comes in a convenient pack of 5.