Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Triton∣r X-100 lysis buffer, pH 7.4

Catalog No. AAJ62289AP
Change view
Click to view available options
Quantity:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62289AP 500mL
AAJ62289AK 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62289AP Supplier Thermo Scientific Chemicals Supplier No. J62289AP
Add to Cart

missing translation for 'validMainframeMsg'

It is widely used non-ionic surfactant for recovery of membrane components under mild non-denaturing conditions. Triton X-100 is a commonly used detergent in laboratories. Triton X-100 is widely used to lyse cells to extract protein or organelles, or to permeabilize the membranes of living cells. Apart from laboratory use, Triton X-100 can be found in several types of cleaning compounds,[8] ranging from heavy-duty industrial products to gentle detergents. It is also a popular ingredient in homemade vinyl record cleaning fluids together with distilled water and isopropyl alcohol. It is a good micellar catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 500mL
Format Liquid
Product Type Cell Lysis Reagent
pH 7.4
Content And Storage 2° to 8°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.