Learn More
Electron Microscopy Sciences Triton X-100, 0.08% Aqueous Solution 500 ML
SDP
Catalog No. 5029533
Encompass_Preferred
F.W. 646.85
Triton X-100 is a low concentration soaking solution in distilled water. It is a non-ionic (neutral pH) solution used to soften dried debris on the edge of the knife prior to recommended cleaning. Also used in whole cell preparations.
Catalog No. 50-295-33 Supplier Electron Microscopy Sciences Supplier No. 22146
F.W. 646.85
Triton X-100 is a low concentration soaking solution in distilled water. It is a non-ionic (neutral pH) solution used to soften dried debris on the edge of the knife prior to recommended cleaning. Also used in whole cell preparations.