Learn More
Electron Microscopy Sciences Triton X-100, 1% Aqueous Sol 500 ML
SDP
Catalog No. 5029534
Encompass_Preferred
Prepared 1% Triton X-100 in Deionized-Distilled water. Used for whole-cell preparations. Marek, L.F., and Kelly, R.O. (1983). A simple technique for the visualization of whole mount cytoskeletons with transmission electron microscopy. Anat. Rec. 207, 365. Pudney, J., and Singer, R.H. (1980). Intracellular filament bundles in whole mounts of chick and human myoblasts extracted with Triton X-100. Tissue cell 12, 595.
Catalog No. 50-295-34 Supplier Electron Microscopy Sciences Supplier No. 22145
Prepared 1% Triton X-100 in Deionized-Distilled water. Used for whole-cell preparations. Marek, L.F., and Kelly, R.O. (1983). A simple technique for the visualization of whole mount cytoskeletons with transmission electron microscopy. Anat. Rec. 207, 365. Pudney, J., and Singer, R.H. (1980). Intracellular filament bundles in whole mounts of chick and human myoblasts extracted with Triton X-100. Tissue cell 12, 595.