Learn More
Sigma Aldrich Fine Chemicals Biosciences Triton X-100 laboratory grade, 9002-93-1, MFCD00128254, 100mL
Catalog No. 501657277
Encompass
Linear Formula t-Oct-C6H4-(OCH2CH2)xOH, x= 9-10, laboratory grade, Synonym: 4-(1,1,3,3-Tetramethylbutyl)phenyl-polyethylene glycol, t-Octylphenoxypolyethoxyethanol, Polyethylene glycol tert-octylphenyl ether
Catalog No. 50-165-7277 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. X100100ML
Linear Formula t-Oct-C6H4-(OCH2CH2)xOH, x= 9-10, laboratory grade, Synonym: 4-(1,1,3,3-Tetramethylbutyl)phenyl-polyethylene glycol, t-Octylphenoxypolyethoxyethanol, Polyethylene glycol tert-octylphenyl ether