Learn More
Research Products International Corp Triton X-100, Scintillation Grade, 1 Liter
SDP
Catalog No. 501978193
Encompass_Preferred
TRITON X-100 is a nonionic, octylphenol ethoxylate surfactant with excellent detergency used in multiple applications including textiles and agrochemicals. Triton is specially processed to eliminate quenching and phosphorescence impurities. It is specifically intended for use in liquid scintillation counting applications.
Catalog No. 50-197-8193 Supplier Research Products International Corp Supplier No. 1110361L
TRITON X-100 is a nonionic, octylphenol ethoxylate surfactant with excellent detergency used in multiple applications including textiles and agrochemicals. Triton is specially processed to eliminate quenching and phosphorescence impurities. It is specifically intended for use in liquid scintillation counting applications.