Learn More
Research Products International Corp Triton X-100, Scintillation Grade, 500 Milliliters
SDP
Catalog No. 50712703
Encompass_Preferred
TRITON X-100 is a nonionic, octylphenol ethoxylate surfactant with excellent detergency used in multiple applications including textiles and agrochemicals. Triton is specially processed to eliminate quenching and phosphorescence impurities. It is specifically intended for use in liquid scintillation counting applications.
Catalog No. 50-712-703 Supplier Research Products International Corp Supplier No. 111036
TRITON X-100 is a nonionic, octylphenol ethoxylate surfactant with excellent detergency used in multiple applications including textiles and agrochemicals. Triton is specially processed to eliminate quenching and phosphorescence impurities. It is specifically intended for use in liquid scintillation counting applications.