Learn More
Andwin Scientific TRITON-X 100 MB GRADE 50ML
SDP
Catalog No. NC9356391
Encompass
TRITON X-100 Detergent Molecular Biology Grade 500ml. Octylphenoxypolyethoxyethanol. Polyethylene Glycol-p-isooctylphenyl Ether. Clear liquid. Non ionic detergent and emulsifier. Purity: >=97% HPLC. Contaminants: DNase protease RNase: none detect, 1/EA
Catalog No. NC9356391 Supplier Andwin Scientific Supplier No. 648466
TRITON X-100 Detergent Molecular Biology Grade 500ml. Octylphenoxypolyethoxyethanol. Polyethylene Glycol-p-isooctylphenyl Ether. Clear liquid. Non ionic detergent and emulsifier. Purity: >=97% HPLC. Contaminants: DNase protease RNase: none detect, 1/EA