Learn More
VWR INTERNATIONAL LLC TRITON X-100 REAGENT GRADE 1L
Catalog No. NC0481054
Encompass
Non-ionic detergent efficiently separate hydrophilic proteins from membrane spanning, hydrophobic proteins without altering biological activity.
Catalog No. NC0481054 Supplier VWR INTERNATIONAL LLC Supplier No. 97062208
Non-ionic detergent efficiently separate hydrophilic proteins from membrane spanning, hydrophobic proteins without altering biological activity.