Learn More
Sigma Aldrich Fine Chemicals Biosciences Triton X-100 reduced, 92046-34-9, MFCD00132505, 25 g
Catalog No. NC1269868
Encompass
Linear Formula: 4-(C8H17)C6H10(OCH2CH2)nOH, n~10, Synonym: Polyoxyethylene (10) isooctylcyclohexyl ether. Non-ionic detergent used to solubilize membrane-bound proteins.
Catalog No. NC1269868 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 28210325G
Linear Formula: 4-(C8H17)C6H10(OCH2CH2)nOH, n~10, Synonym: Polyoxyethylene (10) isooctylcyclohexyl ether. Non-ionic detergent used to solubilize membrane-bound proteins.