Learn More
Perkin Elmer US LLC Triton X-100 Wetting Agent, 100mL
Supplier:  Perkin Elmer US LLC N9300260
Encompass_Preferred
Polyethylene Glycol p-tert-Octylphenyl Ether C22H38O5, C14H21(OCH2CH2)4OH Triton X-100 is a nonionic detergent, 100% active ingredient, which is often used in biochemical applications to solubilize proteins. Triton X-100 has no antimicrobial properties. Triton X-100 is a nonionic surfactant which has a hydrophilic polyethylene oxide group (on average it has 9.5 ethylene oxide units) and a hydrocarbon lipophilic or hydrophobic group. The hydrocarbon group is a 4-(1,1,3,3-tetramethylbutyl)-phenyl group. It is related to the Pluronic range of detergents marketed by BASF. The pluronics are triblock copolymers of ethylene oxide and propylene oxide. The part formed from ethylene oxide is more hydrophilic than the part from propylene oxide. It is very viscous at room temperature and is thus easiest to use after being gently warmed. Triton X Series surfactants are versatile nonionic surfactants recognized for their wetting, detergency, superior hard surface, metal cleaning and excellent em
Catalog No. 50-904-5759