Learn More
Sigma Aldrich Fine Chemicals Biosciences TritonTM X 100 BioXtra
Catalog No. 502908094
Encompass
Triton(TM) X-100 is a common non-ionic surfactant and emulsifier which is often used in biochemical applications to solubilize proteins. It is considered a comparatively mild detergent non-denaturing and is reported in numerous references as a routinely added reagent. It is utilized for lysing cells to extract protein and cellular organelles. It can also permeabilize the living cell membrane for transfection.
Catalog No. 50-290-8094 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T928410L
Triton(TM) X-100 is a common non-ionic surfactant and emulsifier which is often used in biochemical applications to solubilize proteins. It is considered a comparatively mild detergent non-denaturing and is reported in numerous references as a routinely added reagent. It is utilized for lysing cells to extract protein and cellular organelles. It can also permeabilize the living cell membrane for transfection.