Learn More
Sigma Aldrich Fine Chemicals Biosciences TritonTM X 100 peroxide a
Catalog No. 502914357
Encompass
Triton(TM) X-100 is a common non-ionic surfactant and emulsifier which is often used in biochemical applications to solubilize proteins. It is considered a comparatively mild detergent non-denaturing and is reported in numerous references as a routinely added reagent. It is utilized for lysing cells to extract protein and cellular organelles. It can also permeabilize the living cell membrane for transfection.
Catalog No. 50-291-4357 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. X100PC10X5ML
Triton(TM) X-100 is a common non-ionic surfactant and emulsifier which is often used in biochemical applications to solubilize proteins. It is considered a comparatively mild detergent non-denaturing and is reported in numerous references as a routinely added reagent. It is utilized for lysing cells to extract protein and cellular organelles. It can also permeabilize the living cell membrane for transfection.