Learn More
Sigma Aldrich Fine Chemicals Biosciences Triton X-100 reduced, 92046-34-9, MFCD00132505, 25 g
Catalog No. NC0819737
Encompass
Linear Formula: 4-(C8H17)C6H10(OCH2CH2)nOH, n~10, Synonym: Polyoxyethylene (10) isooctylcyclohexyl ether. Triton X-100 reduced, a nonionic surfactant which has a hydrophilic polyethylene oxide chain, is a common laboratory detergent used for applications such as cell lysis and membrane permeabilization as a component of lysis buffers. Triton X-100 is used for the isolation of lipid rafts.
Catalog No. NC0819737 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. X100RS25G
Linear Formula: 4-(C8H17)C6H10(OCH2CH2)nOH, n~10, Synonym: Polyoxyethylene (10) isooctylcyclohexyl ether. Triton X-100 reduced, a nonionic surfactant which has a hydrophilic polyethylene oxide chain, is a common laboratory detergent used for applications such as cell lysis and membrane permeabilization as a component of lysis buffers. Triton X-100 is used for the isolation of lipid rafts.