Learn More
Strem, An Ascensus Company Trityltetra(PFP)borate, 97% 1g
SDP
Catalog No. 5090108773
Encompass
CAS# 136040-19-2. 1g. Trityltetra(pentafluorophenyl)borate, min. 97%. MFCD03426981. Molecular Weight: 922.40. Molecular Formula: (C6H5)3C+(C6F5)4Bae. Color/form: yellow pwdr. Strem# 05-5000. http://www.strem.com/catalog/v/05-5000/
This item is not returnable. View return policy
Catalog No. 50-901-08773 Supplier Strem, An Ascensus Company Supplier No. 0550001G
CAS# 136040-19-2. 1g. Trityltetra(pentafluorophenyl)borate, min. 97%. MFCD03426981. Molecular Weight: 922.40. Molecular Formula: (C6H5)3C+(C6F5)4Bae. Color/form: yellow pwdr. Strem# 05-5000. http://www.strem.com/catalog/v/05-5000/