Learn More
MSC 3M Backer Pad, 13 in, Polyurethane Foam, Low Speed
Catalog No. 502835613
Encompass
The 3M Trizact Backer Pad is a durable polyurethane foam pad with attachment fabric, designed to securely hold abrasive pads for extended periods through multiple use cycles. It is an integral component of floor maintenance systems, specifically intended for polishing applications on various floor types, including stone.
  • Allows for easy attachment of abrasive pads.
  • Holds pads firmly in place.
  • Does not clog or require frequent cleaning.
  • Significantly longer life compared to standard floor pads.
  • Part of the Stone Floor Protection System.
  • Constructed from double-sided polyurethane foam with loop material.
  • Compressible foam helps conform to low spots or uneven floors.
  • Usable on both sides.
  • Designed for low-speed (175-600 RPM) floor cleaning equipment.
Catalog No. 50-283-5613 Supplier MSC Supplier No. 74784463
The 3M Trizact Backer Pad is a durable polyurethane foam pad with attachment fabric, designed to securely hold abrasive pads for extended periods through multiple use cycles. It is an integral component of floor maintenance systems, specifically intended for polishing applications on various floor types, including stone.
  • Allows for easy attachment of abrasive pads.
  • Holds pads firmly in place.
  • Does not clog or require frequent cleaning.
  • Significantly longer life compared to standard floor pads.
  • Part of the Stone Floor Protection System.
  • Constructed from double-sided polyurethane foam with loop material.
  • Compressible foam helps conform to low spots or uneven floors.
  • Usable on both sides.
  • Designed for low-speed (175-600 RPM) floor cleaning equipment.