Learn More
MSC 3M Trizact Backer Pad, 17 in Diameter, Polyurethane Foam, 175-600 RPM
Catalog No. 502836433
Encompass
The Trizact Backer Pad is a low-speed (175-600 RPM) pad with a foam center and loop material on both sides. It is designed to firmly hold Diamond Pads for extended periods through multiple use cycles, providing an improved alternative to traditional floor pads. This backer pad offers a significantly longer life and consistent performance on stone floors like marble, terrazzo, limestone, travertine, and concrete.
  • Loop material firmly holds Diamond Pads for extended periods through multiple use cycles.
  • Compressible foam helps conform to low spots or uneven floors.
  • Usable on both sides.
  • Allows for easy attachment of pads and firmly holds them in place.
  • Does not clog or require frequent cleaning like floor pads.
  • Long life; one backer pad replaces multiple floor pads.
  • Constructed from double-sided polyurethane foam with loop material.
Catalog No. 50-283-6433 Supplier MSC Supplier No. 74783499
The Trizact Backer Pad is a low-speed (175-600 RPM) pad with a foam center and loop material on both sides. It is designed to firmly hold Diamond Pads for extended periods through multiple use cycles, providing an improved alternative to traditional floor pads. This backer pad offers a significantly longer life and consistent performance on stone floors like marble, terrazzo, limestone, travertine, and concrete.
  • Loop material firmly holds Diamond Pads for extended periods through multiple use cycles.
  • Compressible foam helps conform to low spots or uneven floors.
  • Usable on both sides.
  • Allows for easy attachment of pads and firmly holds them in place.
  • Does not clog or require frequent cleaning like floor pads.
  • Long life; one backer pad replaces multiple floor pads.
  • Constructed from double-sided polyurethane foam with loop material.