Learn More
Med Vet International Trizchlor 4, Spray, Conditioner, For Dogs, Cats & Horses, 8oz Bottle
Catalog No. 502827787
Encompass_Preferred
Contains tris-EDTA and 4% chlorhexidine. Useful in the treatment of Gram-positive and Gram-negative organisms, and may be used for resistant bacterial treatment failures and/or resistant bacterial infections (pyoderma, etc.). For topical use in dogs, cats and horses.
Catalog No. 50-282-7787 Supplier Med Vet International Supplier No. PHVTRIZCHLOR4
Contains tris-EDTA and 4% chlorhexidine. Useful in the treatment of Gram-positive and Gram-negative organisms, and may be used for resistant bacterial treatment failures and/or resistant bacterial infections (pyoderma, etc.). For topical use in dogs, cats and horses.