Learn More
Sigma Aldrich Fine Chemicals Biosciences Trizma base, 77-86-1, MFCD00004679, 1 kg
Catalog No. NC0916113
Encompass
Linear Formula: NH2C(CH2OH)3, Molecular Weight: 121.14, BioPerformance Certified, meets EP, USP testing specifications, cell culture tested, ≥99.9% (titration), Synonym: 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol. Tris is an established basimetric standard and buffer used in biochemistry and molecular biology.
Catalog No. NC0916113 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T60661KG
Linear Formula: NH2C(CH2OH)3, Molecular Weight: 121.14, BioPerformance Certified, meets EP, USP testing specifications, cell culture tested, ≥99.9% (titration), Synonym: 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol. Tris is an established basimetric standard and buffer used in biochemistry and molecular biology.