Learn More
Sigma Aldrich Fine Chemicals Biosciences Trizma base, 77-86-1, MFCD00004679, 500 g
Catalog No. NC0632715
Encompass
Linear Formula: NH2C(CH2OH)3, Molecular Weight: 121.14, Synonym: 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol, BioPerformance Certified, meets EP, USP testing specifications, cell culture tested, ≥99.9% (titration). Tris is an established basimetric standard and buffer used in biochemistry and molecular biology.
Catalog No. NC0632715 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T6066500G
Linear Formula: NH2C(CH2OH)3, Molecular Weight: 121.14, Synonym: 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol, BioPerformance Certified, meets EP, USP testing specifications, cell culture tested, ≥99.9% (titration). Tris is an established basimetric standard and buffer used in biochemistry and molecular biology.