Learn More
Sigma Aldrich Fine Chemicals Biosciences Trizma base, 77-86-1, MFCD00004679, 100 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences T1503100G
Encompass
Linear Formula: NH2C(CH2OH)3, Molecular Weight: 121.14, Synonym: 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol. Primary Standard and Buffer, ≥99.9% (titration), crystalline.
Catalog No. NC0761128