Learn More
Sigma Aldrich Fine Chemicals Biosciences Trizma Pre-set crystals, 100 g
Catalog No. NC1224388
Encompass
BioPerformance Certified, pH 8.0, average Mw 141.8, Synonym: Tris crystals. The product is cell culture tested. It is also tested for low endotoxin levels and analyzed for the absence of nucleases and proteases.
Catalog No. NC1224388 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T8443100G
BioPerformance Certified, pH 8.0, average Mw 141.8, Synonym: Tris crystals. The product is cell culture tested. It is also tested for low endotoxin levels and analyzed for the absence of nucleases and proteases.