Learn More
Sigma Aldrich Fine Chemicals Biosciences TrizmaR base Primary Stand25KG
Catalog No. 502914101
Encompass
Tris is an established basimetric standard and buffer used in biochemistry and molecular biology. It may be used by itself as a buffer or as a component of mixed buffer formulations such as Tris-EDTA (TE) buffer Tris-acetate-EDTA (TAE) buffer Tris-borate-EDTA (TBE) buffer etc. It is pure essentially stable relatively non-hygroscopic and has a high equivalent weight.
Catalog No. 50-291-4101 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T150325KG
Tris is an established basimetric standard and buffer used in biochemistry and molecular biology. It may be used by itself as a buffer or as a component of mixed buffer formulations such as Tris-EDTA (TE) buffer Tris-acetate-EDTA (TAE) buffer Tris-borate-EDTA (TBE) buffer etc. It is pure essentially stable relatively non-hygroscopic and has a high equivalent weight.